missing translation for 'onlineSavingsMsg'
Learn More

Orthoboric acid, BAKER, J.T. Baker™

Product Code. 10419580 Shop All J.T.Baker Products
Change view
Click to view available options
:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10419580 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10419580 Supplier JT Baker Supplier No. 0501.1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Laboratory Reagent

Specifications

pH 3.8 to 4.8
Appearance of Solution Passes Test
CAS 10043-35-3
Melting Point 160™C
Boiling Point 185™C (1013 hPa)
Density 1.44 g/cm³ (25°C)
Heavy Metals (as Pb) max. 15 ppm
Identification Passes Test
Assay Percent Range 99.0 to 100.5%
Quantity 1 kg
Molecular Formula BH3O3
MDL Number MFCD00011337
Synonym Boric acid
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
SMILES OB(O)O
IUPAC Name boric acid
Molecular Weight (g/mol) 61.83
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.