missing translation for 'onlineSavingsMsg'
Learn More

ortho Gliclazide, TRC

Product Code. 30394117
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394117 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30394117 Supplier Toronto Research Chemicals Supplier No. TRCG4098801G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material ortho Gliclazide
CAS 1076198-18-9
Purity Grade Notes HPLC
Molecular Formula C15 H21 N3 O3 S
Quantity 1 g
Synonym 1-(Hexahydrocyclopenta[c]pyrrol-2(1H)-yl)-3-[(2-methylphenyl)sulphonyl]urea, Gliclazide Imp. F (EP)
InChI Formula InChI=1S/C15H21N3O3S/c1-11-5-2-3-8-14(11)22(20,21)17-15(19)16-18-9-12-6-4-7-13(12)10-18/h2-3,5,8,12-13H,4,6-7,9-10H2,1H3,(H2,16,17,19)
SMILES Cc1ccccc1S(=O)(=O)NC(=O)NN2CC3CCCC3C2
IUPAC Name 1-(3,3a,4,5,6,6a-hexahydro-1H-cyclopenta[c]pyrrol-2-yl)-3-(2-methylphenyl)sulfonylurea
Molecular Weight (g/mol) 323.41
Formula Weight 323.1304
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.