missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Ortho AP 1867
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Ortho AP 1867 is a selective binding ligand for the single point mutant of FKBP12F36V. Functionalized with a carboxylic acid group in the ortho-position to enable onward chemistry. The position of the carboxylic acid group represents an 'exit vector' allowing modification without interfering with compound binding ability. Ortho AP 1867 is a precursor for the dTAG compounds .
Specifications
Specifications
| Chemical Name or Material | 2-(2-((R)-3-(3,4-Dimethoxyphenyl)-1-(((S)-1-((S)-2-(3,4,5-trimethoxyphenyl)butanoyl)piperidine-2-carbonyl)oxy)propyl)phenoxy)acetic acid |
| CAS | 2230613-03-1 |
| Quantity | 50 mg |
| Target | FKBP12 |
| Molecular Formula | C38H47NO11 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction