missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Orphenadrine citrate, MedChemExpress
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Orphenadrine citrate is a NMDA receptor antagonist with Ki of 6.0 (addition)/- 0.7 (mu)M, HERG potassium channel blocker.
Specifications
Specifications
| Chemical Name or Material | Orphenadrine citrate |
| Physical Form | Solid |
| CAS | 4682-36-4 |
| Color | White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C24H31NO8 |
| For Use With (Application) | Neuroscience-Neurodegeneration |
| Solubility Information | DMSO : 100 mg/mL (216.68 mM; Need ultrasonic) ∣H2O : 10 mg/mL (21.67 mM; Need ultrasonic) |
| SMILES | CC1=CC=CC=C1C(C2=CC=CC=C2)OCCN(C)C.O=C(CC(C(O)=O)(O)CC(O)=O)O |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction