missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ORP9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582972
This item is not returnable.
View return policy
Description
Immunogen sequence: QGLDSGFVPS VQDFDKKLTE ADAYLQILIE QLKLFDDKLQ NCKEDEQRKK IETLKETTNS MVESIKHCIV LLQIAKDQSN AEKHADGM Highest antigen sequence indentity to the following orthologs: Mouse - 93%, Rat - 83%.
This gene encodes a member of a subfamily of LIM domain proteins that are characterized by an N-terminal proline rich region and three C-terminal LIM domains. The encoded protein localizes to the cell periphery in focal adhesions and may be involved in cell-cell adhesion and cell motility. This protein also shuttles through the nucleus and may function as a transcriptional co-activator. This gene is located at the junction of certain disease related chromosomal translocations which result in the expression of chimeric proteins that may promote tumor growth. Alternate splicing results in multiple transcript variants.
Specifications
| ORP9 | |
| Polyclonal | |
| Unconjugated | |
| OSBPL9 | |
| 2600011I06Rik; AU015843; FLJ12492; FLJ14629; FLJ14801; FLJ32055; FLJ34384; MGC15035; Orp9; ORP-9; OSBP4; OSBPL9; OSBP-related protein 9; oxysterol binding protein like 9; oxysterol binding protein-like 9; oxysterol-binding protein-like protein 9; Oxysterol-binding protein-like protein 9-like protein; oxysterol-binding protein-related protein 9; RP4-657D16.1; Unknown (protein for MGC:140169); zgc:154069 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114883 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96SU4 | |
| OSBPL9 | |
| Recombinant protein corresponding to Human OSBPL9. Recombinant protein control fragment (Product #RP-94136). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction