missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORP9 Polyclonal Antibody
ORP9 Polyclonal Antibody
Brand: Invitrogen PA542751
This item is not returnable.
View return policy
Description
ORP9 Polyclonal Antibody for Western Blot, IHC
Specifications
| ORP9 | |
| Polyclonal | |
| Unconjugated | |
| OSBPL9 | |
| 2600011I06Rik; AU015843; FLJ12492; FLJ14629; FLJ14801; FLJ32055; FLJ34384; MGC15035; Orp9; ORP-9; OSBP4; OSBPL9; OSBP-related protein 9; oxysterol binding protein like 9; oxysterol binding protein-like 9; oxysterol-binding protein-like protein 9; Oxysterol-binding protein-like protein 9-like protein; oxysterol-binding protein-related protein 9; RP4-657D16.1; Unknown (protein for MGC:140169); zgc:154069 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 114883 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96SU4 | |
| OSBPL9 | |
| Synthetic peptide directed towards the N-terminal of human OSBPL9 (aa 251-300). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction