missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ORP7 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142079
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Like most members, the encoded protein contains an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain. Two transcript variants encoding the same isoform have been identified.
Specifications
| ORP7 | |
| Polyclonal | |
| Unconjugated | |
| OSBPL7 | |
| ORP7; ORP-7; OSBPL7; OSBP-related protein 7; oxysterol binding protein like 7; oxysterol binding protein-like 7; oxysterol-binding protein-related protein 7 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 114881 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9BZF2 | |
| OSBPL7 | |
| Peptide with sequence C-EPGYGNMDGAVLW. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction