missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ORP5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111096
This item is not returnable.
View return policy
Description
Immunogen sequence: LCGLPASATV HPDQDLFPLN GSSLENDAFS DKSERENPEE SDTETQDHSR KTESGSDQSE TPGAPVRRGT TYVEQVQEEL Highest antigen sequence indentity to the following orthologs: Mouse - 80%, Rat - 80%.
This gene encodes a member of the oxysterol-binding protein family, a group of intracellular lipid receptors that play a key role in the maintenance of cholesterol balance in the body. Most members contain an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain. This gene has been shown to be imprinted, with preferential expression from the maternal allele only in placenta. Transcript variants encoding different isoforms have been identified.
Specifications
| ORP5 | |
| Polyclonal | |
| Unconjugated | |
| OSBPL5 | |
| DKFZp586H1719; FLJ31948; FLJ42929; KIAA1534; OBPH1; ORP5; ORP-5; OSBPL5; OSBP-related protein 5; oxysterol binding protein like 5; oxysterol binding protein-like 5; oxysterol-binding protein homolog 1; oxysterol-binding protein homologue 1; oxysterol-binding protein-related protein 5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114879 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H0X9 | |
| OSBPL5 | |
| Recombinant Protein corresonding to Human ORP5. Recombinant protein control fragment (Product #RP-103339). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction