missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORP150/HSP12A Rabbit anti-Human, Clone: 201, Novus Biologicals™
Rabbit Monoclonal Antibody
£286.00 - £429.00
Specifications
| Antigen | ORP150/HSP12A |
|---|---|
| Clone | 201 |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunoprecipitation 1-4 ul/mg of lysate, Immunohistochemistry-Paraffin 1:100-1:500 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18204867
|
Novus Biologicals
NBP2-89890-50UL |
50 μg |
£286.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18223766
|
Novus Biologicals
NBP2-89890-100UL |
100 μg |
£429.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ORP150/HSP12A Monoclonal specifically detects ORP150/HSP12A in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| ORP150/HSP12A | |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunoprecipitation 1-4 ul/mg of lysate, Immunohistochemistry-Paraffin 1:100-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 150 kDa oxygen-regulated protein, 170 kDa glucose-regulated protein, DKFZp686N08236, FLJ97572, glucose-regulated protein 170, GRP170, GRP-170, HSP12A, hypoxia up-regulated 1, hypoxia up-regulated protein 1, ORP-150, ORP150FLJ94899, oxygen regulated protein (150kD) | |
| Recombinant Human ORP150/HSP12A protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 201 | |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Hypoxia | |
| 0.2 um filtered solution in PBS | |
| 10525 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title