missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORP150/HSP12A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-14113
This item is not returnable.
View return policy
Description
ORP150/HSP12A Polyclonal specifically detects ORP150/HSP12A in Human, Chicken samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ORP150/HSP12A | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Simple Western 1:25, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| 150 kDa oxygen-regulated protein, 170 kDa glucose-regulated protein, DKFZp686N08236, FLJ97572, glucose-regulated protein 170, GRP170, GRP-170, HSP12A, hypoxia up-regulated 1, hypoxia up-regulated protein 1, ORP-150, ORP150FLJ94899, oxygen regulated protein (150kD) | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| HYOU1 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: VLQLINDNTATALSYGVFRRKDINTTAQNIMFYDMGSGSTVCTIVTYQMVKTKEAGMQPQLQIRGVGFDRTLGGLEMELRLRERLAGLFNEQRKGQRAKDVRENPRAMAKLLREANRLKTVLSANADHMAQIEGLMDDVDFKAKVTRV | |
| 0.1 mL | |
| Hypoxia | |
| 10525 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction