missing translation for 'onlineSavingsMsg'
Learn More

ORP150/HSP12A Antibody, DyLight 550, Novus Biologicals™

Product Code. 18366164 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18366164 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18366164 Supplier Novus Biologicals Supplier No. NBP299781R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ORP150/HSP12A Polyclonal specifically detects ORP150/HSP12A in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ORP150/HSP12A
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias 150 kDa oxygen-regulated protein, 170 kDa glucose-regulated protein, DKFZp686N08236, FLJ97572, glucose-regulated protein 170, GRP170, GRP-170, HSP12A, hypoxia up-regulated 1, hypoxia up-regulated protein 1, ORP-150, ORP150FLJ94899, oxygen regulated protein (150kD)
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant ORP150/HSP12A (NP_001124463.1; Met 695-Leu 994).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 10525
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.