missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORP150/HSP12A Antibody (6G7-2H5), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59346AF700
This item is not returnable.
View return policy
Description
ORP150/HSP12A Monoclonal specifically detects ORP150/HSP12A in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| ORP150/HSP12A | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| HYOU1 | |
| Raised against a synthetic peptide of human GRP170 | |
| 100 μL | |
| Primary | |
| Detects 170kDa. | |
| Store at 4C in the dark. | |
| IgG2a |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 6G7-2H5 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| 150 kDa oxygen-regulated protein, 170 kDa glucose-regulated protein, DKFZp686N08236, FLJ97572, glucose-regulated protein 170, GRP170, GRP-170, HSP12A, hypoxia up-regulated 1, hypoxia up-regulated protein 1, ORP-150, ORP150FLJ94899, oxygen regulated protein (150kD) | |
| Mouse | |
| Protein G purified | |
| Hypoxia | |
| 10525 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction