missing translation for 'onlineSavingsMsg'
Learn More

ORP150/HSP12A Antibody (6F7), Novus Biologicals™

Product Code. 18386218 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18386218 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18386218 Supplier Novus Biologicals Supplier No. H00010525M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ORP150/HSP12A Monoclonal antibody specifically detects ORP150/HSP12A in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Microarray
TRUSTED_SUSTAINABILITY

Specifications

Antigen ORP150/HSP12A
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Microarray
Classification Monoclonal
Clone 6F7
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:100 to 1:2000, Immunohistochemistry 1:10 to 1:500, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin 1:10 to 1:500, Microarray
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_006380
Gene Alias 150 kDa oxygen-regulated protein, 170 kDa glucose-regulated protein, DKFZp686N08236, FLJ97572, glucose-regulated protein 170, GRP170, GRP-170, HSP12A, hypoxia up-regulated 1, hypoxia up-regulated protein 1, ORP-150, ORP150FLJ94899, oxygen regulated protein (150kD)
Host Species Mouse
Immunogen HYOU1 (NP_006380, 901 a.a. ~ 999 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. EVQYLLNKAKFTKPRPRPKDKNGTRAEPPLNASASDQGEKVIPPAGQTEDAEPISEPEKVETGSEPGDTEPLELGGPGAEPEQKEQSTGQKRPLKNDEL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 10525
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.