missing translation for 'onlineSavingsMsg'
Learn More

ORP150/HSP12A Antibody (6E3-2C3), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-59345

 View more versions of this product

Product Code. 18263872

  • £427.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ORP150/HSP12A Monoclonal specifically detects ORP150/HSP12A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ORP150/HSP12A
Monoclonal
Unconjugated
PBS pH7.4, 50% glycerol with 0.1% Sodium Azide
HYOU1
Recombinant Full length GRP170 Protein
100 μg
Hypoxia
10525
Human, Mouse, Rat
Purified
Western Blot, Immunocytochemistry, Immunofluorescence
6E3-2C3
Western Blot 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500
150 kDa oxygen-regulated protein, 170 kDa glucose-regulated protein, DKFZp686N08236, FLJ97572, glucose-regulated protein 170, GRP170, GRP-170, HSP12A, hypoxia up-regulated 1, hypoxia up-regulated protein 1, ORP-150, ORP150FLJ94899, oxygen regulated protein (150kD)
Mouse
Protein G purified
RUO
Primary
Detects 170 kDa.
Store at -20C.
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only