missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORP150/HSP12A Antibody (201), HRP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89890H
This item is not returnable.
View return policy
Description
ORP150/HSP12A Monoclonal specifically detects ORP150/HSP12A in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ORP150/HSP12A | |
| Monoclonal | |
| HRP | |
| 150 kDa oxygen-regulated protein, 170 kDa glucose-regulated protein, DKFZp686N08236, FLJ97572, glucose-regulated protein 170, GRP170, GRP-170, HSP12A, hypoxia up-regulated 1, hypoxia up-regulated protein 1, ORP-150, ORP150FLJ94899, oxygen regulated protein (150kD) | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human ORP150/HSP12A (NP_001124463.1; Met 695-Leu 994). | |
| 0.1 mL | |
| Hypoxia | |
| 10525 | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 201 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| No cross-reactivity in ELISA with: Human Fetuin A | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction