missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ORP1 Recombinant Rabbit Monoclonal Antibody (23GB4065), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553685
This item is not returnable.
View return policy
Description
This gene encodes a member of the oxysterol-binding protein family, a group of intracellular lipid receptors. Most members contain an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain, although some members contain only the sterol-binding domain. Transcript variants derived from alternative promoter usage and/or alternative splicing exist; they encode different isoforms.
Specifications
| ORP1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| OSBPL1A | |
| DCDC4A; FLJ10217; FLJ50293; FLJ55454; FLJ79410; G430090F17Rik; Gm753; ORP1; ORP-1; Orp1a; Orp1l; OSBP8; OSBPL1; OSBPL1A; OSBPL1B; OSBP-related protein 1; oxysterol binding protein like 1A; oxysterol binding protein-like 1A; oxysterol binding protein-like 1B; oxysterol-binding protein-related protein 1; oxysterol-binding protein-related protein 1 variant 1; oxysterol-binding protein-related protein 1 variant 2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 114876, 259221, 64291 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4065 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8K4M9, Q91XL9, Q9BXW6 | |
| OSBPL1A | |
| A synthesized peptide derived from human ORP1 (920-950AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction