missing translation for 'onlineSavingsMsg'
Learn More

ORP Standard, +450 mV vs. Ag/AgCl (4M KCl Sat. with AgCl Filling Solution), Ricca

Product Code. 31940648
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31940648 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 31940648 Supplier Ricca Chemical Company Supplier No. R5464545500C
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
  • ORP Standard, 450 mV 500 mL Glass amber

Specifications

Packaging Amber Glass Container
CAS 7732-18-5, 7664-93-9, 7783-85-9, 7783-83-7
Type Redox/ORP Electrode Standard
Quantity 500 mL
Grade ACS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.