missing translation for 'onlineSavingsMsg'
Learn More

Orotidine 5'-Monophosphate Trisodium Salt, TRC

Product Code. 30004455
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004455 5 mg 5mg
30010787 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30004455 Supplier Toronto Research Chemicals Supplier No. TRCO6915255MG
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68244-58-6
Molecular Formula C10 H10 N2 O11 P . 3 Na
Molecular Weight (g/mol) 434.136
Synonym Orotidine, 5'-Phosphate Trisodium Salt, 1,2,3,6-Tetrahydro-2,6-dioxo-3-(5-O-phosphono-β-D-ribofuranosyl)-4-pyrimidinecarboxylic Acid Trisodium Salt;
IUPAC Name trisodium;3-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(phosphonatooxymethyl)tetrahydrofuran-2-yl]-2,6-dioxo-pyrimidine-4-carboxylate
SMILES [Na+].[Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP(=O)([O-])[O-])N2C(=O)NC(=O)C=C2C(=O)[O-]

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C10H13N2O11P.3Na/c13-5-1-3(9(16)17)12(10(18)11-5)8-7(15)6(14)4(23-8)2-22-24(19,20)21;;;/h1,4,6-8,14-15H,2H2,(H,16,17)(H,11,13,18)(H2,19,20,21);;;/q;3*+1/p-3/t4-,6-,7-,8-;;;/m1.../s1
Formula Weight 433.9715
Chemical Name or Material Orotidine 5'-Monophosphate Trisodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.