missing translation for 'onlineSavingsMsg'
Learn More

Orotic acid, anhydrous, MP Biomedicals™

Product Code. 11482582 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11482582 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11482582 Supplier MP Biomedicals Supplier No. 0215179010
 Restricted Item
Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

Specifications

CAS 65-86-1
Color White
Molecular Formula C5H4N2O4
Quantity 10 g
Synonym orotic acid, 6-carboxyuracil, orodin, oropur, orotonin, orotonsan, oroturic, orotyl, vitamin b13, whey factor
InChI Key PXQPEWDEAKTCGB-UHFFFAOYSA-N
SMILES C1=C(NC(=O)NC1=O)C(=O)O
IUPAC Name 2,4-dioxo-1H-pyrimidine-6-carboxylic acid
Molecular Weight (g/mol) 156.097
PubChem CID 967
ChEBI CHEBI:16742
Formula Weight 156.1
Physical Form Crystalline Powder
Chemical Name or Material Orotic acid
Show More Show Less
compliance-icons
Product Identifier
  • Orotic Acid Monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Orotic Acid Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.