missing translation for 'onlineSavingsMsg'
Learn More

Orotic acid, 98%, anhydrous

Product Code. 10705272 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
This item is not returnable. View return policy

Product Code. 10705272

Brand: Thermo Scientific Acros 352881000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 65-86-1 | C5H4N2O4 | 156.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65-86-1
Molecular Formula C5H4N2O4
Molecular Weight (g/mol) 156.1
InChI Key PXQPEWDEAKTCGB-UHFFFAOYSA-N
Synonym orotic acid, 6-carboxyuracil, orodin, oropur, orotonin, orotonsan, oroturic, orotyl, vitamin b13, whey factor
PubChem CID 967
ChEBI CHEBI:16742
IUPAC Name 2,4-dioxo-1H-pyrimidine-6-carboxylic acid
SMILES C1=C(NC(=O)NC1=O)C(=O)O

Specifications

Melting Point >300.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula C4N2H3(O)2COOH
Quantity 100 g
Beilstein 25,253
Merck Index 15,6972
Solubility Information Solubility in water: slightly soluble in water
Formula Weight 156.1
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Orotic acid
Show More Show Less
compliance-icons
Product Identifier
  • Orotic acid, anhydrous, 98%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.