missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ORNT1 Polyclonal Antibody

Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 86%; Rat: 100%; Sheep: 93%.
The SLC25A15 gene encodes the mitochondrial ornithine transporter, which transports ornithine across the inner mitochondrial membrane from the cytosol to the mitochondrial matrix. This is a vital step in the urea cycle, which serves to eliminate toxic ammonium ions from the breakdown of nitrogen.
Specifications
Specifications
| Antigen | ORNT1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | SLC25A15 |
| Gene Accession No. | Q9WVD5 |
| Gene Alias | Ab1-114; D13S327; D630044L02Rik; HHH; MGC137403 protein; mitochondrial ornithine transporter 1; ORC1; ornithine transporter 1; ornithine transporter) member 15; ornithine transporter, mitochondrial 1; Ornt1; SLC25A15; solute carrier family 25 (mitochondrial carrier ornithine transporter), member 15; solute carrier family 25 (mitochondrial carrier; ornithine transporter) member 15; solute carrier family 25 member 15; SP1855 |
| Gene Symbols | SLC25A15 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?