missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ornithine Decarboxylase Rabbit anti-Human, Mouse, Rat, Clone: ODC1/2878R, Novus Biologicals™
Rabbit Monoclonal Antibody
£412.00
Specifications
| Antigen | Ornithine Decarboxylase |
|---|---|
| Clone | ODC1/2878R |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 to 2 μg/mL, Immunohistochemistry-Paraffin 0.25-0.5 μg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
Description
Ornithine Decarboxylase Monoclonal specifically detects Ornithine Decarboxylase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.Specifications
| Ornithine Decarboxylase | |
| 0.2 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication | |
| 10 mM PBS with 0.05% BSA | |
| 4953 | |
| IgG | |
| Protein A or G purified |
| ODC1/2878R | |
| Western Blot 1 to 2 μg/mL, Immunohistochemistry-Paraffin 0.25-0.5 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| ODCEC 4.1.1.17, ornithine decarboxylase, ornithine decarboxylase 1 | |
| Recombinant full-length human Ornithine Decarboxylase protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title