missing translation for 'onlineSavingsMsg'
Learn More

Ornithine Decarboxylase (ODC) Ab-1, Mouse Monoclonal Antibody,Lab Vision™ Epredia™

Take advantage of a quality product that delivers accurate results with the Epredia™ Ornithine Decarboxylase (ODC) Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia MS-464-P1

 View more versions of this product

Product Code. 12681507

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

ODC is the initial and rate-limiting enzyme in the biosynthetic pathway of polyamines and is involved in the conversion of ornithine to putrescine. The biological activity of ODC is rapidly induced in response to virtually all agents known to promote cell proliferation including hormones, drugs, growth factors, mitogens, and tumor promoters. Reportedly, ODC mRNA levels are elevated in lung carcinomas as well as in colon adenomas and carcinomas. ODC activity in colorectal carcinomas is greater than those in adenomas and normal mucosa.

Host Species: Mouse

Clone: MP16-2

Isotype: IgG1

Species Reactivity: Human, Rat, and Mouse. Because epitope of MAb MP16-2 (-IWGPTC-) is present in all known eukaryotic ODC sequences, it should react with ODC from other eukaryotes also.

Epitope: aa 355-360 (-IWGPTC-). It is associated with active site of ODC or a locus in its direct vicinity.

Immunogen: A synthetic hexadecapeptide (KPDEKYYSSSIWGPTC) (aa 345-360) from rat ODC

Molecular Weight: 53kDa

Positive Control: Epithelial cells in normal placenta or prostate CA

Cellular Localization: Cytoplasmic Ab-1 recognizes the native ODC and not the inhibited (labeled with H3-DFMO) enzyme.

Recommended for:

  • Immunohistochemistry (Frozen only)
  • Western Blotting (Not verified)

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ornithine Decarboxylase (ODC) Ab-1
Monoclonal
200μg/mL
Mouse
500 μL
Enzyme
Human, Mouse, Rat
IgG1
Immunohistochemistry (Frozen)
MP16-2
Unconjugated
A synthetic hexadecapeptide (KPDEKYYSSSIWGPTC) (aa345-360) from rat ODC
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.