Learn More
Ornithine Decarboxylase (ODC) Ab-1, Mouse Monoclonal Antibody,Lab Vision™ Epredia™
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: MP16-2
Isotype: IgG1
Species Reactivity: Human, Rat, and Mouse. Because epitope of MAb MP16-2 (-IWGPTC-) is present in all known eukaryotic ODC sequences, it should react with ODC from other eukaryotes also.
Epitope: aa 355-360 (-IWGPTC-). It is associated with active site of ODC or a locus in its direct vicinity.
Immunogen: A synthetic hexadecapeptide (KPDEKYYSSSIWGPTC) (aa 345-360) from rat ODC
Molecular Weight: 53kDa
Positive Control: Epithelial cells in normal placenta or prostate CA
Cellular Localization: Cytoplasmic Ab-1 recognizes the native ODC and not the inhibited (labeled with H3-DFMO) enzyme.
Recommended for:
- Immunohistochemistry (Frozen only)
- Western Blotting (Not verified)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Ornithine Decarboxylase (ODC) Ab-1 |
| Applications | Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | MP16-2 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | A synthetic hexadecapeptide (KPDEKYYSSSIWGPTC) (aa345-360) from rat ODC |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.