missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ornithine Decarboxylase Mouse anti-Human, Mouse, Rat, Clone: rODC1/485, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Ornithine Decarboxylase Monoclonal specifically detects Ornithine Decarboxylase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | Ornithine Decarboxylase |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | rODC1/485 |
| Concentration | 0.2 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.5-1 ug/ml, Immunohistochemistry-Paraffin 0.25-0.5 ug/ml |
| Formulation | 10 mM PBS with 0.05% BSA |
| Gene Alias | ODCEC 4.1.1.17, ornithine decarboxylase, ornithine decarboxylase 1 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?