missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ornithine Decarboxylase Antibody (rODC1/487), DyLight 350, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Ornithine Decarboxylase Monoclonal antibody specifically detects Ornithine Decarboxylase in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
Specifications
| Antigen | Ornithine Decarboxylase |
| Applications | Western Blot, ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Clone | rODC1/487 |
| Conjugate | DyLight 350 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | ODCEC 4.1.1.17, ornithine decarboxylase, ornithine decarboxylase 1 |
| Host Species | Mouse |
| Immunogen | Recombinant full-length human Ornithine Decarboxylase protein (Uniprot: P11926) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?