Learn More
Ornithine Carbamoyltransferase Antibody (CL4046) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-59039PE
Product Code. 30296462
  • £558.00 / 0.10mL
Description
Ornithine Carbamoyltransferase Monoclonal antibody specifically detects Ornithine Carbamoyltransferase in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ornithine Carbamoyltransferase
Monoclonal
PE
EC 2.1.3, EC 2.1.3.3, MGC129967, MGC129968, MGC138856, OCTD, ornithine carbamoyltransferase, ornithine carbamoyltransferase, mitochondrial, Ornithine transcarbamylase, OTCase
This antibody was developed using a recombinant protein derived from P00480, with the exact immunogen sequence remaining proprietary.
0.1 mL
metabolism
5009
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL4046
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified