missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ornithine Carbamoyltransferase Antibody (990421) [Janelia Fluor™ 669], Novus Biologicals™
Shop All Bio Techne ProductsDescription
Ornithine Carbamoyltransferase Monoclonal antibody specifically detects Ornithine Carbamoyltransferase in Human samples. It is validated for Immunohistochemistry
Specifications
Specifications
| Antigen | Ornithine Carbamoyltransferase |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Clone | 990421 |
| Conjugate | Janelia Fluor 669 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 2.1.3, EC 2.1.3.3, MGC129967, MGC129968, MGC138856, OCTD, ornithine carbamoyltransferase, ornithine carbamoyltransferase, mitochondrial, Ornithine transcarbamylase, OTCase |
| Host Species | Mouse |
| Immunogen | Synthetic peptide containing human Ornithine Carbamoyltransferase/OTC, Accession # P00480 |
| Purification Method | Protein A or G purified from cell culture supernatant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?