missing translation for 'onlineSavingsMsg'
Learn More

Ornithine Aspartate (L-Ornithine L-Aspartate), Mikromol™

Product Code. 17988264
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17988264 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17988264 Supplier Mikromol Supplier No. MM1572.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Ornithine Aspartate (L-Ornithine L-Aspartate)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Ornithine L-Aspartate
CAS 3230-94-2
Analyte or Component Names L-Ornithine L-Aspartate
Shipping Condition Room Temperature
Molecular Formula C5 H12 N2 O2 . C4 H7 N O4
Certifications/Compliance ISO 9001
Synonym L-Aspartic acid, compd. with L-ornithine (1:1), Aspartic acid, L-, compd. with L-ornithine (1:1) (8CI), L-Ornithine, L-aspartate (1:1), Ornithine, L-, L-aspartate (1:1) (8CI), Aspartic acid compd. with ornithine, Hepamerz, L-Ornithine L-aspartate, Ormeta, Ornithine aspartate, Orparan, Ruigan, Ornithine Aspartate (L-Ornithine L-Aspartate)
InChI Formula InChI=1S/C5H12N2O2.C4H7NO4/c6-3-1-2-4(7)5(8)9;5-2(4(8)9)1-3(6)7/h4H,1-3,6-7H2,(H,8,9);2H,1,5H2,(H,6,7)(H,8,9)/t4-;2-/m00/s1
SMILES NCCC[C@H](N)C(=O)O.N[C@@H](CC(=O)O)C(=O)O
IUPAC Name (2S)-2-aminobutanedioic acid;(2S)-2,5-diaminopentanoic acid
Molecular Weight (g/mol) 265.26
Quantity 250 mg
Formula Weight 265.1274 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.