missing translation for 'onlineSavingsMsg'
Learn More

Ornidazole, MedChemExpress

Product Code. 16430968
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16430968 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16430968 Supplier Medchem Express Supplier No. HYB0508
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ornidazole(Ro 7-0207) is a 5-nitroimidazole derivative with antiprotozoal and antibacterial properties against anaerobic bacteria.

Ornidazole(Ro 7-0207) is a 5-nitroimidazole derivative with antiprotozoal and antibacterial properties against anaerobic bacteria.

Specifications

Chemical Name or Material Ornidazole
CAS 16773-42-5
Color White
Purity Grade Notes Research
Molecular Formula C7H10ClN3O3
Quantity 5 g
Synonym Ro 7-0207
Solubility Information DMSO : ≥ 100 mg/mL (455.31 mM) ∣H2O : 10 mg/mL (45.53 mM; Need ultrasonic)
SMILES CC1=NC=C([N+]([O-])=O)N1CC(CCl)O
Molecular Weight (g/mol) 219.63
Formula Weight 219.63
Percent Purity 99.74%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.