missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ORMDL1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12829292
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12829292 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12829292 Supplier Invitrogen™ Supplier No. PA521146

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is SK-N-SH cell lysate. PA5-21146 can be used with blocking peptide PEP-1260.

The ORMDL (ORM1-like) family of proteins consist of ORMDL1, ORMDL2 and ORMDL3 that may play roles in -gamma-secretase complex maturation. Localized to the membrane of the endoplasmic reticulum, ORMDLs are widely expressed and are implicated in the expression of Presenilin (PSI) which is involved in Alzheimer's disease (AD). Expression of ORMDL1 is down-regulated in PSI mutations, suggesting a possible role as a therapeutic target for AD. ORMDLs have recently been identified as a potential risk factor for childhood asthma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ORMDL1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene ORMDL1
Gene Accession No. Q921I0, Q9P0S3
Gene Alias Adoplin-1; C730042F17Rik; HSPC202; ORM1-like 1; ORM1-like 1 (S. cerevisiae); ORM1-like protein 1; ORMDL sphingolipid biosynthesis regulator 1; ORMDL1
Gene Symbols ORMDL1
Host Species Rabbit
Immunogen A 16 amino acid peptide of human ORMDL1.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100188936, 227102, 94101
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.