missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORM1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16439-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody recognizes a band about 44 kDa in human plasma which may be due to the glycosylation of ORM1 or the dimer formation of the protein.
ORM1 is an acute phase plasma protein synthesized by the liver. The protein is believed to regulate the interaction between blood cells and endothelial cells, and together with haptoglobin and C reactive protein, also regulates the extravasation of the cells during infection and inflammation. ORM1 is a major acute phase reactant, both in human and in rat. In human plasma, ORM1 (44 kDa) is found at levels of 0.5-1.4 mg/mL. Its concentration in blood plasma is elevated during acute inflammation, such as trauma, maligcies, myocardial infarction, rheumatoid arthritis, bacterial infections, after major surgery, in cases of chronic pain etc., and can be used for the diagnosis of inflammatory conditions.Specifications
| ORM1 | |
| Polyclonal | |
| Unconjugated | |
| ORM1 | |
| AGP 1, AGP A, AGP1, Alpha 1 acid glycoprotein 1, OMD 1, ORM, ORM1, orosomucoid 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18405, 5004 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P02763, Q60590 | |
| Orm1 | |
| ORM1 Fusion Protein Ag9758 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction