missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion Star™ A111 Benchtop pH Meter

Product Code. 15786242
Click to view available options
Type:
pH Meter
pH Meter Bio Kit
pH Meter Standard Kit
ph Meter TRIS Kit
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15786242

Brand: Thermo Scientific™ STARA1117

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Take simple pH, mV and temperature measurements with our benchtop pH meter with easy operation for basic lab testing.

Take simple, routine pH, mV and temperature measurements with the Thermo Scientific™ Orion Star™ A111 Benchtop pH Meter. This meter is an easy-to-use and budget-friendly instrument for basic pH analysis in the lab. Perform up to a 3 point pH calibration with easy recall of calibration points and slope. Quickly navigate setup menus and ensure consistent calibrations using the simple keypad layout and on-screen text prompts. Use the included electrode stand to easily place sensors in samples and simplify storage to minimize breakage.

Features:

  • Large, informative screen clearly shows the main information, including pH or mV reading, temperature, meter mode, calibration information and power source.
  • Thermo Scientific™ AUTO-READ™ locks in the stable reading on your screen and ready indicator alerts when readings are stable.
  • Meter offers up to 3 point pH calibration and automatic recognition of USA/NIST and DIN buffers.
  • Use can easily recall calibration slope data for procedure checks.
  • Nonvolatile memory holds up to 50 data points.
  • Included electrode arm and newly designed probe holder make it easier to maintain and place probes into samples.
  • Plug the meter into almost any power outlet with the included universal power adapter or use four AA batteries (sold separately) to run the meter on DC power.
  • The IP54-rated housing can handle splashes and can be wall-mounted if table space is a problem.

Includes:

Orion STAR: A111 Benchtop pH Meter, Part no. STARA1110, includes electrode arm with redesigned holder and universal power adapter

Orion STAR: A111 Benchtop pH Meter Kit, Part no. STARA1115, includes 9157BNUMD Orion Triode™ 3-in-1 pH/ATC probe; Solution kit (60mL each of pH 4, 7 and 10 buffers; storage solution and cleaning solution); electrode arm with redesigned holder and universal power adapter

Orion STAR: A111 Benchtop pH Meter Kit, Part no. STARA1116, includes GD9156BNWP Orion Green electrode; 927007MD Stainless Steel ATC Probe; electrode arm with redesigned holder and universal power adapter

TRUSTED_SUSTAINABILITY

Specifications

Description Orion Star A111 benchtop pH meter TRIS kit with TRIS-compatible ROSS Ultra pH/ATC electrode
Includes Star A111 pH meter, 8157BNUMD ROSS Ultra epoxy-body pH/ATC electrode, 810199 pH buffer kit, electrode stand, 100-240V universal power adapter
Accuracy (Temperature) ±0.1°C
Accuracy (mV) ±0.2 or ±0.05%, whichever is greater
Accuracy (pH) ±0.01
Data Management Details Internal Data Point Memory
Outputs None
Measurement Modes pH, mV, Relative mV with Temperature
Calibration pH, relative mV (RmV) and temperature
Range (pH) -2 to 16
Resolution (pH) 0.01, 0.1
Range (mV) ±1600.0
Resolution (mV) 0.1mV
Calibration Points 1 to 3
Temperature Range (Metric) -5°C to +105°C
Temperature Range (English) 22.0°F to 221.0°F
Resolution (Temperature) 0.1°C, 0.1°F
Temperature Selection Manual or automatic with ATC temperature probe
Temperature Probe Calibration 1 point temperature offset calibration
Display Type LCD
Memory Non-volatile memory preserves log and settings
Data Points 50
Log Function Type Automatic with Auto-Read Measure Mode, Manual with Continuous Measure Mode
Inputs BNC (pH or ORP electrode), 8 pin MiniDIN (ATC temperature probe)
Certifications/Compliance CE, TUV 3-1, FCC Class A
IP Rating IP54
AC Adapter Universal 50-60 Hz, 100-240 VAC power adapter (Cat. No. 1010003)
Dimensions (L x W x H) 24 cm x 18 cm x 11 cm (9.5 in. x 7.1 in. x 4.3 in.)
Weight (Metric) 0.9 kg
Weight (English) 2 lb.
Warranty 3 Year Meter Warranty
Keypad Simple with highlighted measure key, scroll and shortcut keys
Model Orion Star A111 Benchtop pH/mV/RmV/Temperature Meter
Power Supply Universal AC adapter (included) or 4 AA batteries (sold separately)
Probe Type pH electrode, ORP electrode, ATC temperature probe (includes 8157BNUMD ROSS Ultra Triode pH/ATC electrode)
Languages English
Range (Relative mV) ±1999.9mV
Relative Humidity Range 5 to 85%, non-condensing
Isopotential Point 7.00pH
Stability Smart Stability and Smart Averaging
Temperature (English) Operating 41°F to 113°F (ambient)
Temperature (Metric) Operating 5°C to 45°C (ambient)
Channels 1
Battery Type 4 x AA (sold separately)
Type ph Meter TRIS Kit
Battery Life 2000 hr.
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH210-Safety data sheet available on request.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P273-Avoid release to the environment.
  • P321-Specific treatment (see supplemental fist aid instructions on this label).
Supplemental information
  • MIXTURE LIST-Contains : sodium lauryl sulphate

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.