missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Orion™ Star A and Versa Star Automatic Temperature Compensation/Conductivity Zero Plug
Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
Orion Star A, Versa Star Meters
Unit Size:
Each
Description
The Automatic Temperature Compensation/Conductivity Zero Plug is an accessory for the Orion™ Star A and Versa Star meters.
Features:
- pH Meter Accessories
Specifications
Specifications
| Type | Accessory |
| For Use With (Equipment) | Orion Star A, Versa Star Meters |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction