Learn More
Thermo Scientific™ Orion™ Redox/ORP/Temp Electrodes
Efficiently measure ORP and temperature simultaneously without a separate reference electrode in a single instance with these redox/ORP/temp electrodes.
Brand: Thermo Scientific™ 9180BN
Code : 59
Additional Details : Weight : 0.32000kg
Description
Efficiently measure ORP and temperature simultaneously without a separate reference electrode in a single instance. Thermo Scientific™ Orion™ Redox/ORP/Temp Electrodes measure ORP and temperature using a platinum redox sensor, a silver/silver chloride internal reference system and a built-in thermistor which is adapted for Automatic Temperature Compensation (ATC).
Features:- Available in epoxy triode low maintenance and epoxy refillable triode construction
- Built-in thermistor for Automatic Temperature Compensation (ATC)
- Not designed for amperometric or constant current titrations, such as Karl Fischer analysis
Specifications
Epoxy Refillable Triode | |
Epoxy | |
Orion A+ Series Meters | |
Redox/ORP/Temp Electrode |