Learn More
Thermo Scientific™ Orion™ Pure Water™ pH Buffers and pHISA™ Adjustor
Sample well, rain, distilled/deionized, boiler feed and process waters with this adjustor.
Brand: Thermo Scientific™ 700003
Code : A0
Additional Details : Weight : 0.80000kg
Description
Sample well water, rain water, distilled/deionized water, boiler feed water, process water and more with the Thermo Scientific™ Orion™ Pure Water™ pH Buffers and pHISA™ Adjustor. It offers fast, accurate pH results in high purity and low ionic strength waters.
Features:
- Increases measurement accuracy
- Reduces electrode drift
- Eliminates contamination of sample
Pure Water pH Test Kit:
- Kit contains special low ionic strength buffers for calibration and pHISA™ adjustor additive, to increase sample ionic strength without changing its pH
- For best results, a high quality pH electrode such as the Thermo Scientific™ Orion™ ROSS Ultra™ glass-body pH electrode, Cat. No. 8102BNUWP, is recommended
Specifications
pH Buffer/Ionic Strength Adjustor | |
Clear | |
Provide fast, accurate pH results in high purity and low ionic strength waters. Recommended for samples such as well water, rain water, distilled/deionized water, boiler feed water and process water. | |
4 to 7.1 | |
60 mL | |
Bottle |
100°C | |
pHISA Adjustor Solution Bulk Pack | |
pH Meter and Electrode | |
Solution | |
Potassium Chloride | |
Soluble |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.