missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Orion™ pH Electrode Cleaning Solutions
missing translation for 'orderingAttributeHoverText'
Filling Solution Type:
A, B, C, D
Bacteria Removal (B)
General Cleaning (C)
No Cal Stain Lifter
Oil and Grease Removal (D)
Protein Removal (A)
Rinse
Quantity:
10 pk.
25 pk.
4 x 30 mL
4 x 60 mL
60 mL
60 mL (B, D), 30 mL (A, C)
missing translation for 'unitSize'
60mL
Pack of 10
Pack of 25
Pack of 4
Description
Restore proper electrode performance and prolong electrode life using Thermo Scientific™ Orion™ pH Electrode Cleaning Solutions in ready to use bottles.
Cleaning Solutions:
- Solutions are identified by the letters A, B, C, and D; each letter designates a unique formulation
- Cleaning solution A is for removing protein contaminants, cleaning solution B is for remov
Spécification
Spécification
| Description | 1 Bottle Each of Cleaning Solutions A, B, C, and D; Beaker and Pipet |
| For Use With (Equipment) | pH Electrode |
| Type | Cleaning Solution Set |
| Filling Solution Type | A, B, C, D |
| For Use With (Application) | Solutions restore proper electrode performance and prolong the useful life of the electrode |
| Packaging | Bottle |
| Quantity | 60 mL (B, D), 30 mL (A, C) |
Sécurité et traitement
Identifiant produit
- pH cleaning solution kit
- Danger
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- LONG-TERM AQUATIC HAZARD Chronic 2
- H314-Causes severe skin burns and eye damage.
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
- H411-Toxic to aquatic life with long lasting effects.
- EUH210-Safety data sheet available on request.
- EUH208-Contains . May produce an allergic reaction.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P321-Specific treatment (see on this label).
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P202-Do not handle until all safety precautions have been read and understood.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : hydrochloric acid, sodium hypochlorite,pepsin, methylparaben
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu