missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ pH Electrode Cleaning Solutions

Restore proper electrode performance and prolong electrode life using these pH electrode cleaning solutions.

Brand:  Thermo Scientific™ 900024

55.83 GBP valid until 2024-04-30
Use promo code "21656" to get your promotional price.



Code : A0

Additional Details : Weight : 0.30600kg


 View more versions of this product

Product Code. 11757777

  • £63.68 / Pack of 4
Estimated Shipment: 10-05-2024
to see stock.
Add to basket

Description

Description

Restore proper electrode performance and prolong electrode life using Thermo Scientific™ Orion™ pH Electrode Cleaning Solutions in ready to use bottles.

Cleaning Solutions:

 

  • Solutions are identified by the letters A, B, C, and D; each letter designates a unique formulation
  • Cleaning solution A is for removing protein contaminants, cleaning solution B is for remov
Specifications

Specifications

Solution D 4 x 2 oz. (4 x 60mL) Bottles of Cleaning Solution D for Oil and Grease Removal, Beaker and Pipet
pH Electrode Cleaning Solution
60 mL
pH Electrode
Cleaning
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • pH Cleaner D

Signal Word
  • Warning

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH210-Safety data sheet available on request.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P202-Do not handle until all safety precautions have been read and understood.

Supplemental information
  • MIXTURE LIST-Contains : sodium lauryl sulphate, triton X-100, sodium phosphate dibasic, potassium dihydrogen phosphate, meth

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.