missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ pH Electrode Cleaning Solutions

Product Code. 11747777
Change view
Click to view available options
Filling Solution Type:
A, B, C, D
Bacteria Removal (B)
General Cleaning (C)
No Cal Stain Lifter
Oil and Grease Removal (D)
Protein Removal (A)
Rinse
Quantity:
10 pk.
25 pk.
4 x 30 mL
4 x 60 mL
60 mL
60 mL (B, D), 30 mL (A, C)
Unit Size:
60mL
Pack of 10
Pack of 25
Pack of 4
Product Code. Filling Solution Type Quantity unitSize
11747777 General Cleaning (C) 4 x 30 mL Pack of 4
11722737 No Cal Stain Lifter 60 mL 60mL
11772787 A, B, C, D 60 mL (B, D), 30 mL (A, C) Pack of 4
15304338 Protein Removal (A) 4 x 30 mL Pack of 4
11727777 Protein Removal (A) 4 x 30 mL Pack of 4
11737777 Bacteria Removal (B) 4 x 60 mL Pack of 4
11757777 Oil and Grease Removal (D) 4 x 60 mL Pack of 4
11772168 Rinse 10 pk. Pack of 10
11702188 Rinse 25 pk. Pack of 25
9 options
This item is not returnable. View return policy

Product Code. 11747777

Brand: Thermo Scientific™ 900023

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Restore proper electrode performance and prolong electrode life using these pH electrode cleaning solutions.

Restore proper electrode performance and prolong electrode life using Thermo Scientific™ Orion™ pH Electrode Cleaning Solutions in ready to use bottles.

Cleaning Solutions:

 

  • Solutions are identified by the letters A, B, C, and D; each letter designates a unique formulation
  • Cleaning solution A is for removing protein contaminants, cleaning solution B is for remov
TRUSTED_SUSTAINABILITY

Specifications

Description 4 x 1 oz. (4 x 30mL) Bottles of Cleaning Solution C for General Cleaning, Beaker and Pipet
For Use With (Equipment) pH Electrode
Type Cleaning Solution
Filling Solution Type General Cleaning (C)
For Use With (Application) Solutions restore proper electrode performance and prolong the useful life of the electrode
Packaging Bottle
Quantity 4 x 30 mL
compliance-icons
Product Identifier
  • pH Cleaner C
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P321-Specific treatment (see on this label).
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Hydrochloric Acid

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.