Learn More
Thermo Scientific™ Orion™ pH Buffer Cubitainers™
Hold 5 gallons (19L) of pH buffer in color-coded Thermo Scientific™ Orion™ pH Buffer Cubitainers™ with easy-pour spigot.
Brand: Thermo Scientific™ 9107CB
Code : A0
Additional Details : Weight : 19.00000kg
Description
Hold 5 gallons (19L) of pH buffer in color-coded Thermo Scientific™ Orion™ pH Buffer Cubitainers™ with easy-pour spigot.
- Color coded: pH 4.01 buffer is pink, pH 7.00 buffer is yellow, pH 10.01 buffer is blue
- Packaged with buffer/temperature tables for accurate calibrations and NIST™ traceability
- Manufactured under ISO 9000 quality standards
Specifications
pH Buffer, 5 gallon cubitainers | |
Light Yellow | |
Accurate, reproducible pH calibrations | |
7 | |
19 L | |
Yes |
100°C | |
pH 7.00 Buffer, 5 gallon | |
pH Meter and Electrode | |
Liquid | |
Cubitainer | |
Soluble |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.