Learn More
Thermo Scientific™ Orion™ All-in-One™ pH Buffer Kits
Work with ease using this color-coded pH buffer kit that includes three buffers, storage solution and a storage bottle.
Brand: Thermo Scientific™ 916099
45.53 GBP valid until 2024-05-31
Use promo code "21664" to get your promotional price.
Code : 04
Additional Details : Weight : 0.38430kg
Description
Thermo Scientific™ Orion™ All-in-One™ pH buffer kits include pH 4.01, 7.00 and 10.01 buffers that are color-coded for easy identification as well as storage solution to help ensure your pH electrode is properly maintained. Using high-quality pH buffers to calibrate your pH electrode and pH meter is the best way to ensure your pH measurements are accurate and reproducible. Orion pH buffers are NIST traceable and manufactured under ISO 9001 quality standards.
All Orion pH Buffer Bottles have pH/temperature tables printed on the labels for accurate calibrations and NIST traceability. Manufactured under ISO 9000 quality standards.
Buffer Color Codes:
- pH 4.01 buffer is pink
- pH 5.00 buffer is orange
- pH 7.00 buffer is yellow
- pH 10.01 buffer is blue
The buffers are available in pint (475mL) bottles, portable five packs of 2 oz. (5 x 60mL) bottles, convenient All-in-One buffer kits, 5 gallon (19L) Cubitainers™ and single-use pouches.
Specifications
Small Bottle All-in-One pH Buffer Kit | |
4.01, 7.00, 10.01 | |
Kit | |
60 mL bottle each of pH 4.01, 7.00 and 10.01 buffers; 60 mL bottle of pH electrode storage solution; and 60 mL bottle of pH electrode cleaning solution |
Accurate calibrations and NIST traceability, portable and field analysis | |
5 x 60 mL | |
Yes |
Safety and Handling
- all in one buffer set
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H412-Harmful to aquatic life with long lasting effects.
- EUH210-Safety data sheet available on request.
Precautionary Statement
- P273-Avoid release to the environment.
Supplemental information
- MIXTURE LIST-Contains : sodium lauryl sulphate, triton X-100
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.