missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Orion™ Single Junction Sure-Flow™ Glass Bodied Combination pH Electrode
Description
Thermo Scientific™ Orion™ Sure-Flow electrodes are built with a unique free-flowing liquid-to-liquid junction that simplifies measurements of dirty or viscous samples by helping to prevent clogging, making them easy to clean. They are available in chemically resistant glass bodies and best suited for use in strong acids/bases, pH > 12 or pH < 2, food & cosmetic emulsions, pure water and slurries.
Orion™ Single Junction Electrodes provide a reliable option for routine pH measurement. These silver chloride electrodes are available in a variety of styles and provide accurate measurements of typical samples.
Solutions required
Electrode fill solution
900011 4M KCl with Ag/AgCl filling solution, 5 x 60 mL bottles
Electrode storage solution
910001 pH electrode storage solution, 475 mL bottle
pH buffers
910168 pH 1.68 buffer, NIST traceable, 475 mL
910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
910112 pH 12.46 buffer, NIST traceable, 475 mL
910199 Standard All-in-One pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each, pH
electrode storage solution, 475ml, and 12 mm electrode storage bottle
electrode storage solution, 475ml, and 12 mm electrode storage bottle
pH cleaning solution kit
900020 Cleaning solution A and C, 30 ml each. B and D, 60 ml each
Items included with
900011 4M KCl with Ag/AgCl filling solution 60mL 1 bottle
Warranty period
12 months from date of purchase
Specifications
Specifications
| Description | Model 9172BNWP - Sure-Flow Junction |
| Junction | Sure-Flow |
| Material | Glass Body |
| Type | pH Electrode |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?