Learn More
Thermo Scientific™ Orion™ Lead Electrodes
Measure lead ions in aqueous solutions quickly, simply, accurately and economically with these lead electrodes.
Brand: Thermo Scientific™ 9482SC
Code : 59
Additional Details : Weight : 1.00000kg
Description
Measure lead ions in aqueous solutions quickly, simply, accurately and economically with Thermo Scientific™ Orion™ Lead Electrodes. 9682BNWP is a combination electrode with a Sure-Flow™ junction that allows easy cleaning. When conventional electrode junctions clog, the Sure-Flow junction can be opened and flushed clean simply by pressing the cap. The lead electrode makes a sensitive endpoint detector for the titration of sulfate ions with lead perchlorate.
- Free-flowing Themo Scientific™ Sure-Flow™ junction assures stable, drift-free potentials
- When conventional electrode junctions clog, the Sure-Flow junction can be opened and flushed clean simply by pressing the cap
- Lead Half-Cell Electrode used in conjunction with the Orion 900200 Double Junction Reference electrode offers the benefits of the Sure-Flow™ junction design
- Calibration standards catalog number: 948206 - 0.1 M Pb(CIO4)2; 948207 - 0.1 M Na2SO4
- Required ISA Catalog numbers: Consult instruction manual
Specifications
Screw Cap | |
drift-free potentials | |
ISE Meters |
With screw cap connector | |
Free-flowing Sure-Flow™ junction assures stable | |
Lead Ion Selective Electrode, screw cap connector |