missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ ISE Surfactant Analysis Solutions

Product Code. 11702747
Change view
Click to view available options
Quantity:
60 mL
475 mL
Chemical Name or Material:
Hyamine 1622 Titrant
Sodium Lauryl Sulfate Standard
Surfactant Reagent, Nonionic
Triton X-100 Sample Additive
Unit Size:
475mL
60mL
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Chemical Name or Material unitSize
11702747 475 mL Surfactant Reagent, Nonionic 475mL
12889201 60 mL Sodium Lauryl Sulfate Standard 60mL
12879201 475 mL Hyamine 1622 Titrant 475mL
11792737 475 mL Triton X-100 Sample Additive Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11702747 Supplier Thermo Scientific™ Supplier No. 654205

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use Thermo Scientific™ Orion™ ISE Surfactant Analysis Solutions for titrations with surfactant ion selective electrodes (ISE).

Use Thermo Scientific™ Orion™ ISE Surfactant Analysis Solutions for titrations with surfactant ion selective electrodes (ISE).

All surfactant analysis solutions are packaged as one 1 pint (475mL) bottle, except Cat. No. 654202 packaged as one 2 oz. (60mL) bottle.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Surfactant Reagent, Nonionic
Color Blue
Description 654205
For Use With (Equipment) Surfactant Ion Selective Electrode
Physical Form Liquid
Quantity 475 mL
Solution Type Surfactant Analysis

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.