Learn More
Thermo Scientific™ Orion™ Iodide Ion Selective Electrodes
Product Code. 10288420
Change view
Click to view available options
Connector Types:
BNC
BNC (Waterproof)
Screw cap
Unit Size:
Each
Product Code. Connector Types unitSize
10288420 Screw cap Each
10169860 BNC Each
10191351 BNC (Waterproof) Each
3 options
Product Code. 10288420
Brand: Thermo Scientific™ 9453SC
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Get fast, stable and reliable iodide ions readings in aqueous solutions.
Get fast and stable readings using Thermo Scientific™ Orion™ Iodide Electrodes with built-in Thermo Scientific™ Sure-Flow™ reference. Combination design decreases the amount of required sample and reduces waste.
  • Calibration standards catalog number: 945306 - 0.1M NaI Required ISA catalog number: 940011
TRUSTED_SUSTAINABILITY
Specifications
Connector Types Screw cap
Description With screw cap connector
For Use With (Equipment) ISE Meter
Type Combination ISE