missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ Combination pH Electrodes

Product Code. 11567054
Change view
Click to view available options
Filling Solution Type:
Gel-filled
Liquid-filled
Junction:
Double
Single
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Filling Solution Type Junction unitSize
11567054 Gel-filled Single Each
11547054 Gel-filled Double Each
10359834 Liquid-filled Double Each
10709185 Liquid-filled Single Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11567054 Supplier Thermo Scientific™ Supplier No. GS9106BNWP
  • ONLINE EXCLUSIVE PRICE
    £144.00
    £124.20 / Pack of 1
    Save £19.80
    13% Off
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Environmentally friendly lab pH electrodes to meet RoHS requirements by containing no lead, mercury or other hazardous substances — making disposal of old electrodes easy and hassle free.

Thermo Scientific™ Orion™ Green Electrodes are the first lab electrode to contain no lead, mercury or other hazardous substances and are completely ROHS compliant. GD9156BNWP is a refillable electrode that provides reliable and accurate measurements of routine samples but are best suited for treated effluent, distilled water and wastewater. GD9106BNWP and GS9106BNWP contain a low-maintenance gel that is best suited for field or plant use and educational institutes, where minimal maintenance is required.
 

  • No disposal issues, no disposal cost
  • Choice of single-junction models for clean-water measurements, or double-junction models for dirty-water testing
  • Choice of refillable or low-maintenance gel-filled models
  • Duarable epoxy body
  • pH Range: 0 to 14
  • Dimensions: 120 × 12mm, cap dia. 16mm
  • Waterproof BNC connector, 1m cable; fit any meter with BNC input
Warranty:
  • 12 months from date of purchase (GD9156BNWP)
  • 6 months from date of purchase (GS9106BNWP and GD9106BNWP)

Warranty

  • 12 months from date of purchase (GD9156BNWP)
  • 6 months from date of purchase (GS9106BNWP and GD9106BNWP).

TRUSTED_SUSTAINABILITY

Specifications

Description Low-maintenance gel-filled
Connector Types BNC (Waterproof)
Junction Single
Material Epoxy Body
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 90°C
Type Green Combination pH Electrode, gel-filled
Filling Solution Type Gel-filled
For Use With (Equipment) Orion pH Meter
Length (English) Cable 39.3 in.
Length (Metric) Cable 1 m
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.