Learn More
Thermo Scientific™ Orion™ Calcium Electrodes
Measure calcium in aqueous solutions simply, accurately and economically with these calcium electrodes.
Brand:  Thermo Scientific™ 972001
 View more versions of this product
Product Code. 10599701
  • £259.87 / Each
Estimated Shipment: 14-10-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Measure calcium in aqueous solutions simply, accurately and economically with Thermo Scientific™ Orion™ Calcium Electrodes.
9720BNWP is a combination electrode with a Sure-Flow™ junction. When conventional electrode junctions clog, the Sure-Flow junction can be opened and flushed clean simply by pressing the cap. To be used with module 972001.
A separate reference electrode is unnecessary with this electrode, making it convenient to use with small samples.
Related Products:
  • Calibration standards: 922006 (0.1 M CaCl2); 923206 (100ppm)
  • Required ISA: 932011
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Replacement Module for Combination Calcium Electrode 9720BNWP
Free-flowing Sure-Flow junction assures stable, drift-free potentials
ISE Meters
Combination Ion Selective Electrode