missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ Single Junction Combination pH Electrodes

Product Code. 11752837
Change view
Click to view available options
Connector Types:
BNC
Requires BNC-to-USS Adapter
Junction:
Ceramic
HDPE pin
Wick
Type:
Combination pH Electrode
pH/ATC Combination Electrode
Material:
Epoxy Body
Glass Body
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types Junction Type Material Unit Size
11752837 BNC Wick Combination pH Electrode Epoxy Body Each
11742837 BNC Ceramic Combination pH Electrode Glass Body Each
11762837 Requires BNC-to-USS Adapter HDPE pin pH/ATC Combination Electrode Epoxy Body Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11752837 Supplier Thermo Scientific™ Supplier No. 9146BN
  • ONLINE EXCLUSIVE PRICE
    £80.75
    £79.02 / Pack of 1
    Save £1.73
    2% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Confidently measure your samples with the Orion Silver Chloride pH electrodes.

Thermo Scientific™ Orion™ Silver Chloride pH Electrodes are available in different body materials with either refillable or low maintenance gel options too. The 9142BN is a refillable electrode with chemically resistant glass body best suited for general purpose samples, including solvents, while the 9146BN and 9147BN contain a low-maintenance gel that is best suited for field or plant use, and also educational institutes, where minimal maintenance is preferred. The 9147BN also comes with a built-in ATC.
 

Orion™ Single Junction Electrodes provide a reliable option for routine pH measurement. These silver chloride electrodes are available in a variety of styles and provide accurate measurements of typical samples.

Solutions required

Electrode storage solution

  • 910001 pH electrode storage solution, 475 mL bottle

pH buffers

  • 910168 pH 1.68 buffer, NIST traceable, 475 mL
  • 910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
  • 910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
  • 910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
  • 910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
  • 910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
  • 910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
  • 910112 pH 12.46 buffer, NIST traceable, 475 mL
  • 910199 Standard All-in-One pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each, pH electrode storage solution, 475ml, and 12 mm electrode storage bottle

pH cleaning solution kit

  • 900020 Cleaning solution A and C, 30 mL each. B and D, 60 mL each

Warranty period

  • 6 months from date of purchase
TRUSTED_SUSTAINABILITY

Specifications

Description Low maintenance gel combination with epoxy body and fluted guard
Connector Types BNC
Junction Wick
Material Epoxy Body
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 80°C
Type Combination pH Electrode
For Use With (Equipment) Russell meters
Storage Requirements Store in Orion pH electrode storage solution 910001
Temperature (English) Operating 32°F to 176°F
Tip Type Removeable Bulb Guard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.