missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ AquaPro™ Double Junction Combination pH Electrodes

Product Code. 11772837
Change view
Click to view available options
Connector Types:
BNC (Waterproof)
Mini-DIN (Star)
Material:
Epoxy Body
Glass Body
Type:
Combination pH Electrode
pH/ATC Combination Electrode
Tip Type:
Flat Surface
Semi-micro
Standard
Toughened Bulb
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types Material Type Tip Type unitSize
11772837 BNC (Waterproof) Epoxy Body Combination pH Electrode Standard Each
10636043 BNC (Waterproof) Glass Body Combination pH Electrode Semi-micro Each
11722817 BNC (Waterproof) Glass Body Combination pH Electrode Toughened Bulb Each
10809610 Mini-DIN (Star) Epoxy Body pH/ATC Combination Electrode Standard Each
11772827 BNC (Waterproof) Epoxy Body Combination pH Electrode Semi-micro Each
10308861 BNC (Waterproof) Epoxy Body Combination pH Electrode Flat Surface Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11772837 Supplier Thermo Scientific™ Supplier No. 9156APWP
  • ONLINE EXCLUSIVE PRICE
    £336.00
    £288.00 / Pack of 1
    Save £48.00
    14% Off
Estimated Shipment: 19-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy

Measure the pH of viscous liquid samples quickly, and accurately, with clog-resistant Orion™ AquaPro™ pH electrodes.

Thermo Scientific™ Orion™ AquaPro™ Low Maintenance pH Electrodes provide fast response and extended life with the additional advantages of a double junction design. Their clog resistant, open junctions allow for use in difficult samples, while making post-use cleaning, simple. They are available in a variety of different designs, with the standard types being the 9104APWP & the 9156APWP. These have glass and epoxy bodies respectively and are designed for general lab use. The 9135APWP has a uniquely designed, flat sensing bulb that allows for reliable pH measurement on samples such as paper, textiles, skin, meat, cheese, or in agar gel plates. The 9107APMD contains a built-in temperature sensor that accurately measures pH and temperature with just one electrode, so combines the fast response and open junction design of the Orion™ AquaPro™ Electrode with a built-in ATC.
 

Feature improved electrode life, low maintenance, fast response time and clean junctions. Patented reference system and double-junction design prevent silver ions from contacting sample.
  • Measure 0 to 14pH
  • Temperature range: 0° to 60°C (0° to 140°F)
  • Choice of connectors with 1m (3ft.) cable
TRUSTED_SUSTAINABILITY

Specifications

Description Epoxy Body
Connector Types BNC (Waterproof)
Junction Open
Material Epoxy Body
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 60°C
Type Combination pH Electrode
For Use With (Application) General Purpose, rugged use, lab or field use
For Use With (Equipment) Orion pH Meter
Internal Reference Ag/AgCl Double Junction
pH Relative Accuracy ±0.02
Storage Requirements Always store in pH storage solution, Orion 910001. Either store in a beaker of storage solution or reassemble in the storage bottle the electrode was provided with.
Temperature (English) Operating 32°F to 140°F
Tip Type Standard
Length (English) 4.72 in.
Length (English) Cable 39 in.
Length (Metric) 120 mm
Length (Metric) Cable 1 m
Diameter (English) Body 0.47 in.
Diameter (Metric) Body 12 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.