missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ AQUAfast™ AQ4000 Colorimeter and Accessories

Product Code. 11782168
Change view
Click to view available options
Type:
16 mm vials
AQ4000 Colorimeter
Zero Kit
Unit Size:
Each
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
11782168 16 mm vials Pack of 10
11956069 AQ4000 Colorimeter Each
11757737 Zero Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11782168 Supplier Thermo Scientific™ Supplier No. AC2V16
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Measure common water and wastewater parameters using this colorimeter with 65 pre-programmed methods and auto-test ampoule compatibility.

Measure common colorimetric water and wastewater parameters using the Thermo Scientific™ Orion™ AQUAfast AQ4000 Colorimeter. This instrument has 65 pre-programmed methods, automatic selection of four wavelengths and timer. Use auto-test ampoules with Auto-ID barcodes to simplify sample preparation and tests. Ampoules contain pre-measured liquid reagent and fill with the exact volume of sample. The colorimeter reads the barcode and automatically identifies the parameter and initiates the test. This IP67 waterproof meter records 100 data points and offers an average battery life of 2,500 hours intermittent use.

Features:

  • Identifies the species to be measured
  • Ensures error-free readings at a very affordable price
  • Selects the program and initiates the measurement without the user ever having pushed a button
  • Guides user through the setup and measurement with simple operation
  • Waterproof to IP67 standards

Benefits:

  • User can customize calibration and create completely custom method
  • Up to 12 data points can be used to define custom method
  • User can store maximum of 10 methods in the colorimeter's memory
  • User can datalog 100 points in the field with a time and date tag

Ordering Information:

For use with Orion™ AQUAfast™ AQ4000, Orion™ AQUAfast™ AQ3070, Orion™ AQUAfast™ AQ3700 and discontinued Orion AQ2 series meters.

Note: Uses Thermo Scientific™ Auto-ID™ ampoules, liquid, powder and tablet reagent including many methods accepted for EPA, ISO and other regulatory reporting.

TRUSTED_SUSTAINABILITY

Specifications

Type 16 mm vials
For Use With (Equipment) AQ4000 Colorimeter
Includes 16 mm vials, pk. of 10
Length (English) 0.6 in.
Length (Metric) 16 mm
Quantity 10/Pk.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.